Merck
N6-methyladenosine (m6A) is an endogenous A3 adenosine receptor ligand.
Molecular cell (2021-01-21)
Akiko Ogawa, Chisae Nagiri, Wataru Shihoya, Asuka Inoue, Kouki Kawakami, Suzune Hiratsuka, Junken Aoki, Yasuhiro Ito, Takeo Suzuki, Tsutomu Suzuki, Toshihiro Inoue, Osamu Nureki, Hidenobu Tanihara, Kazuhito Tomizawa, Fan-Yan Wei
ABSTRACT
About 150 post-transcriptional RNA modifications have been identified in all kingdoms of life. During RNA catabolism, most modified nucleosides are resistant to degradation and are released into the extracellular space. In this study, we explored the physiological role of these extracellular modified nucleosides and found that N6-methyladenosine (m6A), widely recognized as an epigenetic mark in RNA, acts as a ligand for the human adenosine A3 receptor, for which it has greater affinity than unmodified adenosine. We used structural modeling to define the amino acids required for specific binding of m6A to the human A3 receptor. We also demonstrated that m6A was dynamically released in response to cytotoxic stimuli and facilitated type I allergy in vivo. Our findings implicate m6A as a signaling molecule capable of activating G protein-coupled receptors (GPCRs) and triggering pathophysiological responses, a previously unreported property of RNA modifications.
Sigma-Aldrich
Cytidine, 99%
Sigma-Aldrich
Albumin, dinitrophenyl, lyophilized powder
Sigma-Aldrich
5-Methylcytidine, ≥99%
Sigma-Aldrich
Uridine, ≥99%
Large Column Holder, For columns o.d. 50 mm, ÄKTA®xpress, Cytiva 28400737
Sigma-Aldrich
Pseudouridine, ≥98% (HPLC)
Sigma-Aldrich
Inosine, ≥99% (HPLC)
Sigma-Aldrich
L-Methionine sulfone
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
7-Methylguanosine, ≥90%
Sigma-Aldrich
Guanosine, ≥98%
Sigma-Aldrich
E-64d, protease inhibitor
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
1,4-Bis(acryloyl)piperazine, BioXtra, suitable for electrophoresis, ≥99.0% (TLC)