Merck
  • Assessment of dopamine D₁ receptor affinity and efficacy of three tetracyclic conformationally-restricted analogs of SKF38393.
Assessment of dopamine D₁ receptor affinity and efficacy of three tetracyclic conformationally-restricted analogs of SKF38393.
Bioorganic & medicinal chemistry (2011-08-25)
Alia H Clark, John D McCorvy, Val J Watts, David E Nichols
ABSTRACT
To assess the effect of conformational mobility on receptor activity, the β-phenyl substituent of dopamine D(1) agonist ligands of the phenylbenzazepine class, (±)-6,6a,7,8,9,13b-hexahydro-5H-benzo[d]naphtho[2,1-b]azepine-11,12-diol (8), and its oxygen and sulfur bioisosteres 9 and 10, respectively, were synthesized as conformationally-restricted analogs of SKF38393, a dopamine D(1)-selective partial agonist. Compounds trans-8b, 9, and 10 showed binding affinity comparable to that of SKF38393, but functionally, they displayed only very weak agonist activity. These results suggest that the conformationally-restricted structure of the analogs cannot adopt a binding orientation that is necessary for agonist activity.
Sigma-Aldrich
R(+)-SCH-23390 hydrochloride, ≥98% (HPLC), solid
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
(R)-(+)-SKF-38393 hydrochloride, ≥98% (HPLC), solid