Merck
  • Activation and inhibition of CTP synthase from Trypanosoma brucei, the causative agent of African sleeping sickness.
Activation and inhibition of CTP synthase from Trypanosoma brucei, the causative agent of African sleeping sickness.
Bioorganic & medicinal chemistry letters (2011-08-16)
Craig H Steeves, Stephen L Bearne
ABSTRACT
CTP Synthase from Trypanosoma brucei (TbCTPS) catalyzes the conversion of UTP to CTP and is a recognized target for the development of antiprotozoal agents. GTP activates glutamine-dependent CTP formation catalyzed by TbCTPS at concentrations below 0.2 mM, but inhibits this activity at concentrations above 0.2 mM. TbCTPS catalyzes ammonia-dependent CTP formation, which is inhibited by purine derivatives such as GTP, guanosine, caffeine, and uric acid with IC(50) values of 460, 380, 480, and 100 μM, respectively. These observations suggest that the purine ring may serve as a useful scaffold for the development of inhibitors of trypanosomal CTP synthase.
Product Description
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Guanosine, BioReagent, suitable for cell culture
Sigma-Aldrich
Guanosine, ≥98%
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Guanosine, ≥97.0% (HPLC)
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg