Merck
  • Cilia density and flow velocity affect alignment of motile cilia from brain cells.
Cilia density and flow velocity affect alignment of motile cilia from brain cells.
The Journal of experimental biology (2020-12-31)
Nicola Pellicciotta, Debasish Das, Jurij Kotar, Marion Faucourt, Nathalie Spassky, Eric Lauga, Pietro Cicuta
ABSTRACT
In many organs, thousands of microscopic 'motile cilia' beat in a coordinated fashion generating fluid flow. Physiologically, these flows are important in both development and homeostasis of ciliated tissues. Combining experiments and simulations, we studied how cilia from brain tissue align their beating direction. We subjected cilia to a broad range of shear stresses, similar to the fluid flow that cilia themselves generate, in a microfluidic setup. In contrast to previous studies, we found that cilia from mouse ependyma respond and align to these physiological shear stress at all maturation stages. Cilia align more easily earlier in maturation, and we correlated this property with the increase in multiciliated cell density during maturation. Our numerical simulations show that cilia in densely packed clusters are hydrodynamically screened from the external flow, in agreement with our experimental observation. Cilia carpets create a hydrodynamic screening that reduces the susceptibility of individual cilia to external flows.
Product Description
Corning® Transwell® polyester membrane cell culture inserts, 24 mm Transwell with 0.4 μm pore polyester membrane insert, TC-treated, sterile, 24/cs
Pricing and availability is not currently available.
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt ≥300,000
Pricing and availability is not currently available.