Merck
  • Helminth-induced regulation of T-cell transfer colitis requires intact and regulated T cell Stat6 signaling in mice.
Helminth-induced regulation of T-cell transfer colitis requires intact and regulated T cell Stat6 signaling in mice.
European journal of immunology (2020-10-18)
Ahmed Metwali, Sarah Winckler, Joseph F Urban, Mark H Kaplan, M Nedim Ince, David E Elliott
ABSTRACT
Infection with parasitic worms (helminths) alters host immune responses and can inhibit pathogenic inflammation. Helminth infection promotes a strong Th2 and T regulatory response while suppressing Th1 and Th17 function. Th2 responses are largely dependent on transcriptional programs directed by Stat6-signaling. We examined the importance of intact T cell Stat6 signaling on helminth-induced suppression of murine colitis that results from T cell transfer into immune-deficient mice. Colonization with the intestinal nematode Heligmosomoides polygyrus bakeri resolves WT T cell transfer colitis. However, if the transferred T cells lack intact Stat6 then helminth exposure failed to attenuate colitis or suppress MLN T cell IFN-γ or IL17 production. Loss of Stat6 signaling resulted in decreased IL10 and increased IFN-γ co-expression by IL-17+ T cells. We also transferred T cells from mice with constitutive T cell expression of activated Stat6 (Stat6VT). These mice developed a severe eosinophilic colitis that also was not attenuated by helminth infection. These results show that T cell expression of intact but regulated Stat6 signaling is required for helminth infection-associated regulation of pathogenic intestinal inflammation.
Sigma-Aldrich
2,3,4,5,6-Pentafluorothiophenol, 97%
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by gel-filtration chromatography