Merck
  • Influence of the addition of different ingredients on the bioaccessibility of glucose released from rice during dynamic in vitro gastrointestinal digestion.
Influence of the addition of different ingredients on the bioaccessibility of glucose released from rice during dynamic in vitro gastrointestinal digestion.
International journal of food sciences and nutrition (2020-05-19)
Jean-Michel Fernandes, Daniel Alexandre Madalena, António Augusto Vicente, Ana Cristina Pinheiro
ABSTRACT
Rice represents a primary source of carbohydrates in human nutrition. Upon its consumption, the released sugars are mostly absorbed, categorising rice as a high glycemic index food. Addition of ingredients is common practice when cooking rice, which may affect rice digestibility and influence nutrients absorption in the gastrointestinal (GI) tract, enabling a controlled glucose release. In this sense, rice formulations were submitted to a dynamic in vitro GI model, constituted by reactors that simulates peristalsis coupled to filtration membranes, to evaluate carbohydrates hydrolysis and bioaccessibility. Addition of quinoa and wholegrains reduced carbohydrates hydrolysis (i.e. 38.5 ± 5.08% and 57.98 ± 1.91%, respectively) and glucose bioaccessibility (i.e. 25.92 ± 5.70% and 42.56 ± 1.39%, respectively) when compared with brown rice (i.e. 63.86 ± 2.96% hydrolysed and 44.33 ± 1.88% absorbed). Addition of vegetables significantly decreased sample chewiness and resulted in superior hydrolysis (71.75 ± 7.44%) and glucose absorption (51.61 ± 6.25%).
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥2,500 units/mg protein (E1%/280)
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Ethyl pyruvate-1-13C, ≥99 atom % 13C, ≥98% (CP)
Sigma-Aldrich
Pancreatin from porcine pancreas, 8 × USP specifications
Sigma-Aldrich
Bile extract porcine