Merck
  • Carbonic anhydrase inhibitors. Inhibition of the β-class enzymes from the fungal pathogens Candida albicans and Cryptococcus neoformans with branched aliphatic/aromatic carboxylates and their derivatives.
Carbonic anhydrase inhibitors. Inhibition of the β-class enzymes from the fungal pathogens Candida albicans and Cryptococcus neoformans with branched aliphatic/aromatic carboxylates and their derivatives.
Bioorganic & medicinal chemistry letters (2011-03-16)
Fabrizio Carta, Alessio Innocenti, Rebecca A Hall, Fritz A Mühlschlegel, Andrea Scozzafava, Claudiu T Supuran
ABSTRACT
The inhibition of the β-carbonic anhydrases (CAs, EC 4.2.1.1) from the pathogenic fungi Cryptococcus neoformans (Can2) and Candida albicans (Nce103) with a series of 25 branched aliphatic and aromatic carboxylates has been investigated. Human isoforms hCA I and II were also included in the study for comparison. Aliphatic carboxylates were generally millimolar hCA I and II inhibitors and low micromolar/submicromolar β-CA inhibitors. Aromatic carboxylates were micromolar inhibitors of the four enzymes but some of them showed low nanomolar activity against the fungal pathogenic enzymes. 4-Hydroxy- and 4-methoxy-benzoate inhibited Can2 with K(I)s of 9.5-9.9 nM. The methyl esters, hydroxamates, hydrazides and carboxamides of some of these derivatives were also effective inhibitors of the α- and β-CAs investigated here.
Product Description
Supelco
Melting point standard 182-184°C, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Methyl p-anisate, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Aminobenzoic acid sodium salt, ≥99%, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium propionate, ≥99.0%
Pricing and availability is not currently available.
Supelco
Sodium 4-hydroxybenzoate, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
p-Anisic acid, ≥99%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium propionate, FCC
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Methyl p-anisate, ≥99%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium propionate, meets analytical specification of NF, 99-100.5% (ex dried subst.)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium propionate, ≥99.0%, BioReagent, suitable for insect cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Pricing and availability is not currently available.
Sigma-Aldrich
2-Ethylhexanoic acid, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium formate, BioUltra, ≥99.0% (NT)
Pricing and availability is not currently available.
Supelco
2-Ethylhexanoic acid, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium benzoate, puriss., meets analytical specification of Ph. Eur., BP, FCC, E211, 99.0-100.5% (calc. to the dried substance), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium butyrate, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Methoxybenzoic acid, ReagentPlus®, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium benzoate, purum p.a., ≥99.0% (NT)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium 2-ethylhexanoate, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium benzoate, ReagentPlus®, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Ethylhexanoic acid, ≥99%
Pricing and availability is not currently available.