Merck
  • A microbial metabolite remodels the gut-liver axis following bariatric surgery.
A microbial metabolite remodels the gut-liver axis following bariatric surgery.
Cell host & microbe (2021-01-13)
Snehal N Chaudhari, James N Luo, David A Harris, Hassan Aliakbarian, Lina Yao, Donggi Paik, Renuka Subramaniam, Arijit A Adhikari, Ashley H Vernon, Ayse Kiliç, Scott T Weiss, Jun R Huh, Eric G Sheu, A Sloan Devlin
ABSTRACT
Bariatric surgery is the most effective treatment for type 2 diabetes and is associated with changes in gut metabolites. Previous work uncovered a gut-restricted TGR5 agonist with anti-diabetic properties-cholic acid-7-sulfate (CA7S)-that is elevated following sleeve gastrectomy (SG). Here, we elucidate a microbiome-dependent pathway by which SG increases CA7S production. We show that a microbial metabolite, lithocholic acid (LCA), is increased in murine portal veins post-SG and by activating the vitamin D receptor, induces hepatic mSult2A1/hSULT2A expression to drive CA7S production. An SG-induced shift in the microbiome increases gut expression of the bile acid transporters Asbt and Ostα, which in turn facilitate selective transport of LCA across the gut epithelium. Cecal microbiota transplant from SG animals is sufficient to recreate the pathway in germ-free (GF) animals. Activation of this gut-liver pathway leads to CA7S synthesis and GLP-1 secretion, causally connecting a microbial metabolite with the improvement of diabetic phenotypes.
Sigma-Aldrich
DL-Cysteine, technical grade
Pricing and availability is not currently available.
Sigma-Aldrich
Ampicillin sodium salt, powder or crystals, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human NR1I2
Pricing and availability is not currently available.
Sigma-Aldrich
Neomycin trisulfate salt hydrate, meets USP testing specifications, powder
Pricing and availability is not currently available.
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, from non-animal source, suitable for cell culture, meets EP, USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Golgicide A, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human NR1H4
Pricing and availability is not currently available.