Merck
  • NRG/ErbB signaling regulates neonatal muscle growth but not neuromuscular contractures in neonatal brachial plexus injury.
NRG/ErbB signaling regulates neonatal muscle growth but not neuromuscular contractures in neonatal brachial plexus injury.
FEBS letters (2021-01-10)
Brendan L Ho, Qingnian Goh, Sia Nikolaou, Liangjun Hu, Kritton Shay-Winkler, Roger Cornwall
ABSTRACT
Neonatal brachial plexus injury (NBPI) causes disabling and incurable muscle contractures that are driven by impaired growth of denervated muscles. A rare form of NBPI, which maintains afferent muscle innervation despite motor denervation, does not cause contractures. As afferent innervation regulates various aspects of skeletal muscle homeostasis through NRG/ErbB signaling, our current study investigated the role of this pathway in modulating contracture development. Through pharmacologic modification with an ErbB antagonist and NRG1 isoforms, we discovered that NRG/ErbB signaling does not modulate the development of contractures in neonatal mice. Instead, ErbB inhibition impeded growth in nondenervated skeletal muscles, whereas increased ErbB activation exacerbated denervation-induced skeletal muscle atrophy. This potential regulatory effect of NRG/ErbB signaling on neonatal muscle growth warrants deeper investigation.
Sigma-Aldrich
Anti-phospho-ERBB2 (pTyr1248) antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.