Merck
  • 17-DMAG dually inhibits Hsp90 and histone lysine demethylases in alveolar rhabdomyosarcoma.
17-DMAG dually inhibits Hsp90 and histone lysine demethylases in alveolar rhabdomyosarcoma.
iScience (2021-01-26)
Shivendra Singh, Ahmed Abu-Zaid, Wenwei Lin, Jonathan Low, Alireza Abdolvahabi, Hongjian Jin, Qiong Wu, Bailey Cooke, Jie Fang, John Bowling, Sivaraja Vaithiyalingam, Duane Currier, Mi-Kyung Yun, Dinesh M Fernando, Julie Maier, Heather Tillman, Purva Bulsara, Zhaohua Lu, Sourav Das, Anang Shelat, Zhenmei Li, Brandon Young, Richard Lee, Zoran Rankovic, Andrew J Murphy, Stephen W White, Andrew M Davidoff, Taosheng Chen, Jun Yang
ABSTRACT
Histone lysine demethylases (KDMs) play critical roles in oncogenesis and therefore may be effective targets for anticancer therapy. Using a time-resolved fluorescence resonance energy transfer demethylation screen assay, in combination with multiple orthogonal validation approaches, we identified geldanamycin and its analog 17-DMAG as KDM inhibitors. In addition, we found that these Hsp90 inhibitors increase degradation of the alveolar rhabdomyosarcoma (aRMS) driver oncoprotein PAX3-FOXO1 and induce the repressive epigenetic mark H3K9me3 and H3K36me3 at genomic loci of PAX3-FOXO1 targets. We found that as monotherapy 17-DMAG significantly inhibits expression of PAX3-FOXO1 target genes and multiple oncogenic pathways, induces a muscle differentiation signature, delays tumor growth and extends survival in aRMS xenograft mouse models. The combination of 17-DMAG with conventional chemotherapy significantly enhances therapeutic efficacy, indicating that targeting KDM in combination with chemotherapy may serve as a therapeutic approach to PAX3-FOXO1-positive aRMS.
Sigma-Aldrich
2,3-Dihydroxybenzoic acid, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 units
1 Estimated to ship on 09 March 2025
US$150.00
200 units
1 Estimated to ship on 09 March 2025
US$283.00
500 units
1 Estimated to ship on 09 March 2025
US$479.00
1000 units
1 Estimated to ship on 09 March 2025
US$779.00
2000 units
1 Estimated to ship on 09 March 2025
US$1,400.00
Sigma-Aldrich
GSK-J4, ≥98% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 units
1 Estimated to ship on 09 March 2025
US$150.00
200 units
1 Estimated to ship on 09 March 2025
US$283.00
500 units
1 Estimated to ship on 09 March 2025
US$479.00
1000 units
1 Estimated to ship on 09 March 2025
US$779.00
2000 units
1 Estimated to ship on 09 March 2025
US$1,400.00
Sigma-Aldrich
17-(Allylamino)-17-demethoxygeldanamycin, ≥98% (HPLC), solid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 units
1 Estimated to ship on 09 March 2025
US$150.00
200 units
1 Estimated to ship on 09 March 2025
US$283.00
500 units
1 Estimated to ship on 09 March 2025
US$479.00
1000 units
1 Estimated to ship on 09 March 2025
US$779.00
2000 units
1 Estimated to ship on 09 March 2025
US$1,400.00
Sigma-Aldrich
Poly(ethylene glycol), average Mn 300
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 units
1 Estimated to ship on 09 March 2025
US$150.00
200 units
1 Estimated to ship on 09 March 2025
US$283.00
500 units
1 Estimated to ship on 09 March 2025
US$479.00
1000 units
1 Estimated to ship on 09 March 2025
US$779.00
2000 units
1 Estimated to ship on 09 March 2025
US$1,400.00
Sigma-Aldrich
TWEEN® 80, suitable for cell culture, suitable for insect cell culture, viscous liquid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 units
1 Estimated to ship on 09 March 2025
US$150.00
200 units
1 Estimated to ship on 09 March 2025
US$283.00
500 units
1 Estimated to ship on 09 March 2025
US$479.00
1000 units
1 Estimated to ship on 09 March 2025
US$779.00
2000 units
1 Estimated to ship on 09 March 2025
US$1,400.00
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 units
1 Estimated to ship on 09 March 2025
US$150.00
200 units
1 Estimated to ship on 09 March 2025
US$283.00
500 units
1 Estimated to ship on 09 March 2025
US$479.00
1000 units
1 Estimated to ship on 09 March 2025
US$779.00
2000 units
1 Estimated to ship on 09 March 2025
US$1,400.00