Merck
  • KDM2B promotes IL-6 production and inflammatory responses through Brg1-mediated chromatin remodeling.
KDM2B promotes IL-6 production and inflammatory responses through Brg1-mediated chromatin remodeling.
Cellular & molecular immunology (2019-06-15)
Qingqing Zhou, Yunkai Zhang, Bo Wang, Wenhui Zhou, Yong Bi, Wanwan Huai, Xi Chen, Yihan Chen, Zhongmin Liu, Xingguang Liu, Zhenzhen Zhan
ABSTRACT
IL-6 plays important and pleiotropic roles in infection and inflammatory diseases, and its production needs to be tightly regulated. However, the epigenetic mechanism underlying Il6 gene transcription remains to be fully elucidated. Here, we report that lysine-specific demethylase 2b (KDM2B), which demethylates H3K4me3 and H3K36me2, is required in macrophages and dendritic cells for the induction of IL-6 but not TNF-α, IL-1, and IFN-β. Compared to wild-type mice, KDM2B-deficient mice were more resistant to endotoxin shock and colitis, with a less severe inflammatory pathogenesis phenotype and decreased IL-6 production in sera. KDM2B selectively bound the Il6 promoter but did not alter histone demethylation; instead, KDM2B interacted with Brahma-related gene 1 (Brg1), the core ATPase subunit of SWI/SNF chromatin remodeling complexes, to facilitate chromatin accessibility of the Il6 promoter. Furthermore, KDM2B directly recruited RNA Polymerase II to further initiate and promote Il6 transcription. Thus, our finding identifies a novel nonclassical function of KDM2B in gene-specific transcription initiation and enhancement of Il6 independent of its demethylase activity and adds new insight into the specific epigenetic modification mechanism of inflammatory immune responses.
Sigma-Aldrich
Anti-JHDM1B Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
ChIPAb+ Dimethyl-Histone H3 (Lys36) - ChIP Validated Antibody and Primer Set, serum, from rabbit
Sigma-Aldrich
ChIPAb+ Trimethyl-Histone H3 (Lys4) - ChIP Validated Antibody and Primer Set, rabbit monoclonal, from rabbit