Merck
  • The cytotoxic properties and preferential toxicity to tumour cells displayed by some 2,4-bis(benzylidene)-8-methyl-8-azabicyclo[3.2.1] octan-3-ones and 3,5-bis(benzylidene)-1-methyl-4-piperidones.
The cytotoxic properties and preferential toxicity to tumour cells displayed by some 2,4-bis(benzylidene)-8-methyl-8-azabicyclo[3.2.1] octan-3-ones and 3,5-bis(benzylidene)-1-methyl-4-piperidones.
European journal of medicinal chemistry (2008-05-13)
Hari N Pati, Umashankar Das, Swagatika Das, Brian Bandy, Erik De Clercq, Jan Balzarini, Masami Kawase, Hiroshi Sakagami, J Wilson Quail, James P Stables, Jonathan R Dimmock
ABSTRACT
This study demonstrated that replacement of the axial protons on the C2 and C6 atoms of various 1-methyl-3,5-bis(benzylidene)-4-piperidones 3 by a dimethylene bridge leading to series 2 lowered cytotoxic potencies. Four compounds 2a and 3a-c emerged as lead molecules based on their toxicity towards different neoplasms and their selective toxicity for malignant rather than normal cells. Some possible reasons for the disparity between the IC(50) values in the two series of compounds are presented based on molecular modeling, logP values and respiration in rat liver mitochondria.
Sigma-Aldrich
3-Octanone, ≥98%
Supelco
3-Octanone, analytical standard
Sigma-Aldrich
3-Octanone, ≥98%, FG
Sigma-Aldrich
Melphalan, powder