Merck
  • Molecular modeling and inhibition of phospholipase A2 by polyhydroxy phenolic compounds.
Molecular modeling and inhibition of phospholipase A2 by polyhydroxy phenolic compounds.
European journal of medicinal chemistry (2008-04-22)
S L da Silva, A K Calgarotto, V Maso, D C S Damico, P Baldasso, C L Veber, J A F P Villar, A R M Oliveira, M Comar, K M T Oliveira, S Marangoni
ABSTRACT
Phospholipases A(2) are enzymes responsible for the hydrolysis of membrane phospholipids that release arachidonic acid, which serves as substrate for pro-inflammatory mediators, such as prostaglandins and leucotriens. The design of specific inhibitors for PLA(2) might help in the development of new anti-inflammatory drugs. Polyhydroxy phenolic compounds, such as flavonoids, vitamin E, rosmarinic acid and aristolochic acid, are able to inhibit PLA(2) from different sources. Herein, we have studied the kinetic behavior and the capacity of inhibiting edema formation induced by PLA(2) of five different polyhydroxy phenolic compounds (two phenolic derivatives and three acetophenone hydroxylated derivatives) extracted from the venom of Crotalus adamanteus. The results showed that compounds 1,3-dihydroxy benzene, 1,3,5-trihydroxy benzene and 2,4,6-trihydroxy acetophenone were the most efficient in the inhibition of the enzymatic activity and edema induction by PLA(2). It was also verified that the number of hydroxyls in each molecule is not a limiting factor for the inhibition capacity of these compounds. Molecular modeling studies indicated that the most active compounds are linked to the amino acid Asp 49 and that they destabilize the coordination of the calcium atom, which is essential to the catalytic activity. The study of potential surfaces showed that there are conditions in which the potential values must be adequate for enzyme complex formation with polyhydroxy phenolic compounds. When the potential over the hydroxyl surfaces is very high, formation of stable complexes does not occur and the enzyme does not act intensely. These results might be helpful in the design of a drug that specifically inhibits PLA(2).
Sigma-Aldrich
Resorcinol, BioXtra, ≥99%
Sigma-Aldrich
2′,6′-Dihydroxyacetophenone, 97%
Sigma-Aldrich
Resorcinol, ACS reagent, ≥99.0%
Sigma-Aldrich
Resorcinol, ≥98%, FG
Sigma-Aldrich
Resorcinol, ReagentPlus®, 99%
Sigma-Aldrich
Phloroglucinol, Used to detect the presence of wood fiber.
Sigma-Aldrich
Phloroglucinol, suitable for plant cell culture, BioReagent
Sigma-Aldrich
Phloroglucinol, ≥99.0% (HPLC)