Merck
  • Nontoxic Cobalt(III) Schiff Base Complexes with Broad-Spectrum Antifungal Activity.
Nontoxic Cobalt(III) Schiff Base Complexes with Broad-Spectrum Antifungal Activity.
Chemistry (Weinheim an der Bergstrasse, Germany) (2020-11-25)
Angelo Frei, A Paden King, Gabrielle J Lowe, Amy K Cain, Francesca L Short, Hue Dinh, Alysha G Elliott, Johannes Zuegg, Justin J Wilson, Mark A T Blaskovich
ABSTRACT
Resistance to currently available antifungal drugs has quietly been on the rise but overshadowed by the alarming spread of antibacterial resistance. There is a striking lack of attention to the threat of drug-resistant fungal infections, with only a handful of new drugs currently in development. Given that metal complexes have proven to be useful new chemotypes in the fight against diseases such as cancer, malaria, and bacterial infections, it is reasonable to explore their possible utility in treating fungal infections. Herein we report a series of cobalt(III) Schiff base complexes with broad-spectrum antifungal activity. Some of these complexes show minimum inhibitory concentrations (MIC) in the low micro- to nanomolar range against a series of Candida and Cryptococcus yeasts. Additionally, we demonstrate that these compounds show no cytotoxicity against both bacterial and human cells. Finally, we report the first in vivo toxicity data on these compounds in Galleria mellonella, showing that doses as high as 266 mg kg-1 are tolerated without adverse effects, paving the way for further in vivo studies of these complexes.
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder
Sigma-Aldrich
Resazurin sodium salt, powder, BioReagent
Corning® 384 well plates, lid, sterile; γ-irradiated
Sigma-Aldrich
Anidulafungin, ≥97% (HPLC)
Deepwell 96 well plate, polypropylene, sterile, pack of 24 ea
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Triton X-100, Molecular Biology
Sigma-Aldrich
Melittin from honey bee venom, ≥85% (HPLC)