Merck
  • Reconsideration of the Semaphorin-3A Binding Motif Found in Chondroitin Sulfate Using Galnac4s-6st-Knockout Mice.
Reconsideration of the Semaphorin-3A Binding Motif Found in Chondroitin Sulfate Using Galnac4s-6st-Knockout Mice.
Biomolecules (2020-11-05)
Satomi Nadanaka, Shinji Miyata, Bai Yaqiang, Jun-Ichi Tamura, Osami Habuchi, Hiroshi Kitagawa
ABSTRACT
The chondroitin sulfate (CS)-rich dense extracellular matrix surrounding neuron cell bodies and proximal dendrites in a mesh-like structure is called a perineuronal net (PNN). CS chains in PNNs control neuronal plasticity by binding to PNN effectors, semaphorin-3A (Sema3A) and orthodenticle homeobox 2. Sema3A recognizes CS-containing type-E disaccharide units (sulfated at O-4 and O-6 of N-acetylgalactosamine). Type-E disaccharide units are synthesized by N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase (GalNAc4S-6ST). In this study, we demonstrated that Sema3A accumulates in the PNNs surrounding parvalbumin cells, even in mice deficient in GalNAc4S-6ST. In addition, there were no differences in the number and structure of PNNs visualized by Cat316 antibody and Wisteria floribunda lectin, which recognize CS chains, between wild type and GalNAc4S-6ST knockout mice. Therefore, we re-examined the Sema3A binding motif found in CS chains using chemically synthesized CS tetrasaccharides. As a result, we found that non-sulfated GalNAc residues at the non-reducing termini of CS chains are required for the binding of Sema3A.
Sigma-Aldrich
Anti-Aggrecan Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Chondroitin Sulfate Proteoglycan Antibody, carbohydrate epitope, clone Cat-316, ascites fluid, clone Cat-316, Chemicon®