Merck
  • p53-mediated senescence impairs the apoptotic response to chemotherapy and clinical outcome in breast cancer.
p53-mediated senescence impairs the apoptotic response to chemotherapy and clinical outcome in breast cancer.
Cancer cell (2012-06-16)
James G Jackson, Vinod Pant, Qin Li, Leslie L Chang, Alfonso Quintás-Cardama, Daniel Garza, Omid Tavana, Peirong Yang, Taghi Manshouri, Yi Li, Adel K El-Naggar, Guillermina Lozano
ABSTRACT
Studies on the role of TP53 mutation in breast cancer response to chemotherapy are conflicting. Here, we show that, contrary to dogma, MMTV-Wnt1 mammary tumors with mutant p53 exhibited a superior clinical response compared to tumors with wild-type p53. Doxorubicin-treated p53 mutant tumors failed to arrest proliferation, leading to abnormal mitoses and cell death, whereas p53 wild-type tumors arrested, avoiding mitotic catastrophe. Senescent tumor cells persisted, secreting senescence-associated cytokines exhibiting autocrine/paracrine activity and mitogenic potential. Wild-type p53 still mediated arrest and inhibited drug response even in the context of heterozygous p53 point mutations or absence of p21. Thus, we show that wild-type p53 activity hinders chemotherapy response and demonstrate the need to reassess the paradigm for p53 in cancer therapy.
Sigma-Aldrich
Anti-p53 (Ab-2) (Pantropic) Mouse mAb (PAb1801), liquid, clone PAb1801, Calbiochem®