Merck
  • Unc-13 homolog D mediates an antiviral effect of the chromosome 19 microRNA cluster miR-517a.
Unc-13 homolog D mediates an antiviral effect of the chromosome 19 microRNA cluster miR-517a.
Journal of cell science (2020-10-24)
Kamil Krawczynski, Yingshi Ouyang, Jean-Francois Mouillet, Tianjiao Chu, Carolyn B Coyne, Yoel Sadovsky
ABSTRACT
The function of microRNAs (miRNAs) can be cell autonomous or communicated to other cell types and has been implicated in diverse biological processes. We previously demonstrated that miR-517a-3p (miR-517a), a highly expressed member of the chromosome 19 miRNA cluster (C19MC) that is transcribed almost exclusively in human trophoblasts, attenuates viral replication via induction of autophagy in non-trophoblastic recipient cells. However, the molecular mechanisms underlying these effects remain unknown. Here, we identified unc-13 homolog D (UNC13D) as a direct, autophagy-related gene target of miR-517a, leading to repression of UNC13D. In line with the antiviral activity of miR-517a, silencing UNC13D suppressed replication of vesicular stomatitis virus (VSV), whereas overexpression of UNC13D increased VSV levels, suggesting a role for UNC13D silencing in the antiviral activity of miR-517a. We also found that miR-517a activated NF-κB signaling in HEK-293XL cells expressing TLR8, but the effect was not specific to C19MC miRNA. Taken together, our results define mechanistic pathways that link C19MC miRNA with inhibition of viral replication.
Sigma-Aldrich
CL264, ≥98% (HPLC)
Sigma-Aldrich
TL8-506, ≥98% (HPLC)
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Anti-Ago2 Antibody, clone 11A9, clone 11A9, from rat