Merck
  • Identification and characterization of novel sirtuin inhibitor scaffolds.
Identification and characterization of novel sirtuin inhibitor scaffolds.
Bioorganic & medicinal chemistry (2009-09-08)
Brandi D Sanders, Brittany Jackson, Michael Brent, Alexander M Taylor, Weiwei Dang, Shelley L Berger, Stuart L Schreiber, Konrad Howitz, Ronen Marmorstein
ABSTRACT
The sirtuin proteins are broadly conserved NAD(+)-dependent deacetylases that are implicated in diverse biological processes including DNA recombination and repair, transcriptional silencing, longevity, apoptosis, axonal protection, insulin signaling, and fat mobilization. Because of these associations, the identification of small molecule sirtuin modulators has been of significant interest. Here we report on high throughput screening against the yeast sirtuin, Hst2, leading to the identification of four unique inhibitor scaffolds that also inhibit the human sirtuins, SIRT1-3, and are able to inhibit telomeric silencing of yeast Sir2 in vivo. The identified inhibitor scaffolds range in potency from IC(50) values of 6.5-130 microM against Hst2. Each of the inhibitor scaffolds binds reversibly to the enzyme, and kinetic analysis reveals that each of the inhibitors is non-competitive with respect to both acetyl-lysine and NAD(+) binding. Limited SAR analysis of the scaffolds also identifies which functional groups may be important for inhibition. These sirtuin inhibitors are low molecular weight and well-suited for lead molecule optimization, making them useful chemical probes to study the mechanism and biological roles of sirtuins and potential starting points for optimization into therapeutics.
Sigma-Aldrich
4-Benzylphenol, 99%
Supelco
Hexachlorophene, PESTANAL®, analytical standard
Sigma-Aldrich
N-(2-Bromoethyl)phthalimide, 95%
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)