Merck
  • Design, synthesis and cruzain docking of 3-(4-substituted-aryl)-1,2,4-oxadiazole-N-acylhydrazones as anti-Trypanosoma cruzi agents.
Design, synthesis and cruzain docking of 3-(4-substituted-aryl)-1,2,4-oxadiazole-N-acylhydrazones as anti-Trypanosoma cruzi agents.
Bioorganic & medicinal chemistry (2009-08-18)
José Mauricio dos Santos Filho, Ana Cristina Lima Leite, Boaz Galdino de Oliveira, Diogo Rodrigo Magalhães Moreira, Milena S Lima, Milena Botelho Pereira Soares, Lucia Fernanda C C Leite
ABSTRACT
Research in recent years has demonstrated that the Trypanosoma cruzi cysteine protease cruzain (TCC) is a valid chemotherapeutic target, since inhibitors of this protease affect the pathology appropriately. By exploring the N-acylhydrazones (NAH) as privileged structures usually present in antiparasitic agents, we investigated a library of 16 NAH bearing the 3-(4-substituted-aryl)-1,2,4-oxadiazole scaffold (NAH 3a-h, 4a-h). The in vitro bioactivity against epimastigote and trypomastigote forms of T. cruzi was evaluated, and some NAH under study exhibited antitrypanosomal activity at concentrations that are not toxic to mammalian cells. The series of compounds based on the 3-(4-substituted-aryl)-1,2,4-oxadiazole scaffold revealed the remarkable importance of each substituent at the phenyl's 4-position for the inhibitory activity. Non-nitrated compounds 3a and 4e were found to be as potent as the reference drug, Benznidazole. In addition, the molecular origin of the antitrypanosomal properties for these series was investigated using docking studies of the TCC structure.
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Crystal Violet, indicator for the determination of the redox potential, S. No.: 785
Sigma-Aldrich
Crystal Violet, for microscopy (Bact., Bot., Hist., Vit.), indicator (pH 0.1-2.0)
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Gentian violet, for microscopy (Bact., Hist.)
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission