Merck
  • Potential antioxidants and tyrosinase inhibitors from synthetic polyphenolic deoxybenzoins.
Potential antioxidants and tyrosinase inhibitors from synthetic polyphenolic deoxybenzoins.
Bioorganic & medicinal chemistry (2009-06-02)
Lean-Teik Ng, Horng-Huey Ko, Tzy-Ming Lu
ABSTRACT
Deoxybenzoins (DOBs) are one-pot synthetic precursors of isoflavones with feature analogous to those beneficial polyphenols such as resveratrol (stilbene) and phloretin (dihydrochalcone). In this study, seventeen polyphenolic DOBs were synthesized and evaluated by various antioxidant assays and tyrosinase inhibitory effect in vitro. Results displayed that these DOBs are powerful antioxidants; for example, 2,3,4-trihydroxy-3',4'-dimethoxydeoxybenzoin possesses an excellent anti-lipid peroxidation activity (IC(50)=0.72+/-0.16 microM), whilst 2,4,4',5-tetrahydroxydeoxybenzoin showed good DPPH radical scavenging activity (IC(50)=0.69+/-0.04 microM), which were better than Trolox and vitamin C. Besides exhibiting a weak metal chelating effect, these DOBs were effective in scavenging ABTS(+) and superoxide anion (O2-) radicals. DOBs also exhibited potent mushroom tyrosinase inhibitory activity; for example 2,3,4'-trihydroxy-4-methoxydeoxybenzoin displayed stable and significant inhibitory effect on tyrosinase activity, with IC(50) values 43.37, 43.10 and 46.10 microM at incubation intervals of 0.5, 1.5, and 2.5h, respectively. These results suggest that, with the advantage of being readily synthesizable small molecules, DOBs can be potentially developed into clinical and industrial antioxidants.
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, 99%
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Kojic acid
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis