Merck
  • Antibacterial alkoxybenzamide inhibitors of the essential bacterial cell division protein FtsZ.
Antibacterial alkoxybenzamide inhibitors of the essential bacterial cell division protein FtsZ.
Bioorganic & medicinal chemistry letters (2008-12-10)
Lloyd G Czaplewski, Ian Collins, E Andrew Boyd, David Brown, Stephen P East, Mihaly Gardiner, Rowena Fletcher, David J Haydon, Vincent Henstock, Peter Ingram, Clare Jones, Caterina Noula, Leanne Kennison, Chris Rockley, Valerie Rose, Helena B Thomaides-Brears, Rebecca Ure, Mark Whittaker, Neil R Stokes
ABSTRACT
3-Methoxybenzamide is a weak inhibitor of the essential bacterial cell division protein FtsZ. Exploration of the structure-activity relationships of 3-methoxybenzamide analogues led to the identification of potent anti-staphylococcal compounds.
Sigma-Aldrich
3-Methoxybenzamide, 97%
Sigma-Aldrich
3-Methoxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Mupirocin, ≥92% (HPLC), powder
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Supelco
Ciprofloxacin, VETRANAL®, analytical standard