Merck
  • Characterization of alpha-nitromethyl ketone as a new zinc-binding group based on structural analysis of its complex with carboxypeptidase A.
Characterization of alpha-nitromethyl ketone as a new zinc-binding group based on structural analysis of its complex with carboxypeptidase A.
Bioorganic & medicinal chemistry letters (2009-08-04)
Shou-Feng Wang, Guan Rong Tian, Wen-Zheng Zhang, Jing-Yi Jin
ABSTRACT
Zinc-binding groups (ZBGs) are exhaustively applied in the development of the new inhibitors against a wide variety of physiologically and pathologically important zinc proteases. Here the alpha-nitro ketone was presented as a new ZBG, which is a transition-state analog featured by the unique bifurcated hydrogen bonds at the active site of carboxypeptidase A based on the structural analysis. Introduction of a nitro group at the alpha-position of the ketone could provide more non-covalent interactions without loss of the abilities to form a tetrahedral transition-state analog.
Product Description
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Bromoacetic acid, ReagentPlus®, ≥99.0%
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Bromoacetic acid, reagent grade, 97%
Sigma-Aldrich
Acetic acid, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC