Merck
  • SPARC promotes pancreatic cancer cell proliferation and migration through autocrine secretion into the extracellular milieu.
SPARC promotes pancreatic cancer cell proliferation and migration through autocrine secretion into the extracellular milieu.
Oncology letters (2021-05-11)
Kehua Pan, Xince Huang, Xiufen Jia
ABSTRACT
SPARC is a secreted glycoprotein that plays a complex and multifaceted role in tumour formation and progression. However, whether SPARC is an oncogene or a tumour suppressor is still unclear. Moreover, SPARC demonstrates potential in clinical pancreatic adenocarcinoma (PAAD) treatment, although it has been identified as an oncogene in some studies and a tumor suppressor in others. In the present study, a pan-cancer analysis of SPARC was carried out using The Cancer genome Atlas data, which demonstrated that SPARC was an oncogene in most cancer types and a cancer suppressor in others. In addition, SPARC expression was significantly upregulated in PAAD and associated with poor prognosis. SPARC also promoted the proliferation and migration of PANC-1 and SW1990 cell lines in vitro. SPARC was detected in the culture supernatant of PAAD cells and pancreatic acinar AR42J cells. SPARC regulated PAAD cell proliferation only when secreted into the extracellular milieu, thus explaining why the prognosis of patients with PAAD is correlated with the SPARC expression of both tumour cells and stromal cells. Collectively, the present findings demonstrated that the function of SPARC was associated with tumour type and that SPARC may represent an important oncogene in PAAD that merits further study.
Sigma-Aldrich
Anti-Ī²-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rabbit IgG (H+L), highly cross adsorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
ANTI-FLAGĀ® antibody, Rat monoclonal, clone 6F7, purified from hybridoma cell culture