Merck
  • Immunoglobulin class and subclass distribution of antibodies reactive with the immunodominant antigen of Actinobacillus actinomycetemcomitans serotype b.
Immunoglobulin class and subclass distribution of antibodies reactive with the immunodominant antigen of Actinobacillus actinomycetemcomitans serotype b.
Infection and immunity (1993-06-01)
H Lu, J V Califano, H A Schenkein, J G Tew
ABSTRACT
The aims of this study were to determine the immunodominant antigens of Actinobacillus actinomycetemcomitans serotype b (Aab) for the different immunoglobulin (Ig) classes and subclasses and to determine the relative levels of these different Igs in serum. Seropositive early-onset periodontitis patients were sampled, and the Ig classes IgG, IgA, and IgM and subclasses IgG1, IgG2, IgG3, IgG4, IgA1, and IgA2 were studied. Reactivity with Aab antigens was assessed by using the Western blot (immunoblot) in limiting dilution analysis and radioimmunoassay with sera from 13 early-onset periodontitis subjects. A smeared antigen in the upper portion of the immunoblots, typical of high-molecular-weight LPS, was immunodominant for IgG, IgA, IgM, IgG1, IgG2, IgG3, IgA1, and IgA2. This smeared antigen was present in every patient for all of these Igs at the endpoint. A few additional antigens were also present at the endpoint in some patients, but none were present in more than half of the subjects. The distribution of antibody titers by Ig classes reactive with the Aab immunodominant antigen was IgG > IgA > IgM. The distribution of antibody titers by IgG subclass was IgG2 > IgG1 approximately IgG3. Further quantitation by radioimmunoassay revealed that the mean concentration of IgG2 (65.7 micrograms/ml) was significantly greater than that of IgG1 (8.8 micrograms/ml). The IgA subclass distribution was IgA1 >> IgA2, with IgA1 apparently being second only to IgG2. Therefore, the Aab antigen eliciting the highest antibody level in virtually all Ig classes and subclasses appeared to be lipopolysaccharide, and IgG2 was markedly elevated over all other serum Ig classes or subclasses reactive with Aab.
Sigma-Aldrich
Mouse Anti-Human IgG₄, Fc Fragment Specific (HP6025), liquid, clone HP6025, Calbiochem®