Merck
Heat stress activates YAP/TAZ to induce the heat shock transcriptome.
Nature cell biology (2020-11-18)
Min Luo, Zhipeng Meng, Toshiro Moroishi, Kimberly C Lin, Guobo Shen, Fei Mo, Bin Shao, Xiawei Wei, Ping Zhang, Yuquan Wei, Kun-Liang Guan
ABSTRACT
The Hippo pathway plays critical roles in cell growth, differentiation, organ development and tissue homeostasis, whereas its dysregulation can lead to tumorigenesis. YAP and TAZ are transcription co-activators and represent the main downstream effectors of the Hippo pathway. Here, we show that heat stress induces a strong and rapid YAP dephosphorylation and activation. The effect of heat shock on YAP is dominant to other signals known to modulate the Hippo pathway. Heat shock inhibits LATS kinase by promoting HSP90-dependent LATS interaction with and inactivation by protein phosphatase 5. Heat shock also induces LATS ubiquitination and degradation. YAP and TAZ are crucial for cellular heat shock responses, including the heat shock transcriptome and cell viability. This study uncovers previously unknown mechanisms of Hippo regulation by heat shock, as well as physiological functions of YAP, in the heat stress response. Our observations also reveal a potential combinational therapy involving hyperthermia and targeting of the Hippo pathway.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Glutathione-S-transferase (GST) antibody, Mouse monoclonal, clone 2H3-D10, purified from hybridoma cell culture
Sigma-Aldrich
MG-132, A cell-permeable, potent, reversible proteasome inhibitor (Ki = 4 nM).
Sigma-Aldrich
Copper(II) sulfate hydrate, 98%
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
3-(Biphenyl-4-yl)-5-(4-tert-butylphenyl)-4-phenyl-4H-1,2,4-triazole, 97%