Merck
  • In vitro capture and characterization of embryonic rosette-stage pluripotency between naive and primed states.
In vitro capture and characterization of embryonic rosette-stage pluripotency between naive and primed states.
Nature cell biology (2020-05-06)
Alex Neagu, Emiel van Genderen, Irene Escudero, Lucas Verwegen, Dorota Kurek, Johannes Lehmann, Jente Stel, René A M Dirks, Guido van Mierlo, Alex Maas, Cindy Eleveld, Yang Ge, Alexander T den Dekker, Rutger W W Brouwer, Wilfred F J van IJcken, Miha Modic, Micha Drukker, Joop H Jansen, Nicolas C Rivron, Esther B Baart, Hendrik Marks, Derk Ten Berge
ABSTRACT
Following implantation, the naive pluripotent epiblast of the mouse blastocyst generates a rosette, undergoes lumenogenesis and forms the primed pluripotent egg cylinder, which is able to generate the embryonic tissues. How pluripotency progression and morphogenesis are linked and whether intermediate pluripotent states exist remain controversial. We identify here a rosette pluripotent state defined by the co-expression of naive factors with the transcription factor OTX2. Downregulation of blastocyst WNT signals drives the transition into rosette pluripotency by inducing OTX2. The rosette then activates MEK signals that induce lumenogenesis and drive progression to primed pluripotency. Consequently, combined WNT and MEK inhibition supports rosette-like stem cells, a self-renewing naive-primed intermediate. Rosette-like stem cells erase constitutive heterochromatin marks and display a primed chromatin landscape, with bivalently marked primed pluripotency genes. Nonetheless, WNT induces reversion to naive pluripotency. The rosette is therefore a reversible pluripotent intermediate whereby control over both pluripotency progression and morphogenesis pivots from WNT to MEK signals.
Sigma-Aldrich
Alkaline Phosphatase Detection Kit, This Alkaline Phosphatase Detection Kit is a specific & sensitive tool for the phenotypic assessment of Embryonic Stem (ES) cell differentiation by the determination of AP activity.
Sigma-Aldrich
Wnt Antagonist II, IWP-2, The Wnt Antagonist II, IWP-2, also referenced under CAS 686770-61-6, controls the biological activity of Wnt. This small molecule/inhibitor is primarily used for Cancer applications.