MilliporeSigma
A domino effect in antifolate drug action in Escherichia coli.
Nature chemical biology (2008-08-30)
Yun Kyung Kwon, Wenyun Lu, Eugene Melamud, Nurussaba Khanam, Andrew Bognar, Joshua D Rabinowitz
ABSTRACT
Mass spectrometry technologies for measurement of cellular metabolism are opening new avenues to explore drug activity. Trimethoprim is an antibiotic that inhibits bacterial dihydrofolate reductase (DHFR). Kinetic flux profiling with (15)N-labeled ammonia in Escherichia coli reveals that trimethoprim leads to blockade not only of DHFR but also of another critical enzyme of folate metabolism: folylpoly-gamma-glutamate synthetase (FP-gamma-GS). Inhibition of FP-gamma-GS is not directly due to trimethoprim. Instead, it arises from accumulation of DHFR's substrate dihydrofolate, which we show is a potent FP-gamma-GS inhibitor. Thus, owing to the inherent connectivity of the metabolic network, falling DHFR activity leads to falling FP-gamma-GS activity in a domino-like cascade. This cascade results in complex folate dynamics, and its incorporation in a computational model of folate metabolism recapitulates the dynamics observed experimentally. These results highlight the potential for quantitative analysis of cellular metabolism to reveal mechanisms of drug action.
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dihydrofolic acid, ≥90%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trimethoprim, ≥98.5%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Trimethoprim, VETRANAL®, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Folic acid, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Folic acid, ≥97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%