Merck
  • Hepatic Premalignant Alterations Triggered by Human Nephrotoxin Aristolochic Acid I in Canines.
Hepatic Premalignant Alterations Triggered by Human Nephrotoxin Aristolochic Acid I in Canines.
Cancer prevention research (Philadelphia, Pa.) (2016-02-07)
Ke Jin, Kun-kai Su, Tong Li, Xia-qing Zhu, Qi Wang, Ren-shan Ge, Zong-fu Pan, Bo-wen Wu, Li-jun Ge, Yi-han Zhang, Yi-fan Wang, Guo-fang Shen, Dan-yan Zhu, Chun-sheng Xiang, Lan-juan Li, Yi-jia Lou
ABSTRACT
Aristolochic acid I (AAI) existing in plant drugs from Aristolochia species is an environmental human carcinogen associated with urothelial cancer. Although gene association network analysis demonstrated gene expression profile changes in the liver of human TP53 knock-in mice after acute AAI exposure, to date, whether AAI causes hepatic tumorigenesis is still not confirmed. Here, we show that hepatic premalignant alterations appeared in canines after a 10-day AAI oral administration (3 mg/kg/day). We observed c-Myc oncoprotein and oncofetal RNA-binding protein Lin28B overexpressions accompanied by cancer progenitor-like cell formation in the liver by AAI exposure. Meanwhile, we found that forkhead box O1 (FOXO1) was robustly phosphorylated, thereby shuttling into the cytoplasm of hepatocytes. Furthermore, utilizing microarray and qRT-PCR analysis, we confirmed that microRNA expression significantly dysregulated in the liver treated with AAI. Among them, we particularly focused on the members in let-7 miRNAs and miR-23a clusters, the downstream of c-Myc and IL6 receptor (IL6R) signaling pathway linking the premalignant alteration. Strikingly, when IL6 was added in vitro, IL6R/NF-κB signaling activation contributed to the increase of FOXO1 phosphorylation by the let-7b inhibitor. Therefore, it highlights the new insight into the interplay of the network in hepatic tumorigenesis by AAI exposure, and also suggests that anti-premalignant therapy may be crucial for preventing AAI-induced hepatocarcinogenesis.
Sigma-Aldrich
Anti-Fodrin Mouse mAb (Fod009), liquid, clone Fod009, Calbiochem®
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection