MilliporeSigma
  • The mechanism and role of intracellular α-ketoglutarate reduction in hepatic stellate cell activation.
The mechanism and role of intracellular α-ketoglutarate reduction in hepatic stellate cell activation.
Bioscience reports (2020-03-04)
Jianjian Zhao, Yueping Jiang, Xueguo Sun, Xishuang Liu, Fuguo Liu, Mingquan Song, Lingyun Zhang
ABSTRACT
The activation of hepatic stellate cells (HSCs) plays a central role in liver fibrosis. α-ketoglutarate is a natural metabolite and previous studies have shown that increase in intracellular α-ketoglutarate can inhibit HSC activation. The aim of the present study is to determine the changes and role of intracellular α-ketoglutarate in HSC activation and clarify its mechanism of action. A human HSC cell line (LX-2) and the primary mouse HSC were used in the present study. We detected the changes of intracellular α-ketoglutarate levels and the expression of enzymes involved in the metabolic processes during HSC activation. We used siRNA to determine the role of intracellular α-ketoglutarate in HSC activation and elucidate the mechanism of the metabolic changes. Our results demonstrated that intracellular α-ketoglutarate levels decreased with an HSC cell line and primary mouse HSC activation, as well as the expression of isocitrate dehydrogenase 2 (IDH2), an enzyme that catalyzes the production of α-ketoglutarate. In addition, knockdown of IDH2 efficiently promoted the activation of HSCs, which was able to be reversed by introduction of an α-ketoglutarate analogue. Furthermore, we demonstrated that α-ketoglutarate regulated HSC activation is independent of transforming growth factor-β1 (TGF-β1). Our findings demonstrated that decrease in IDH2 expression limits the production of α-ketoglutarate during HSC activation and in turn promotes the activation of HSCs through a TGF-β1 independent pathway. The present study suggests that IDH2 and α-ketoglutarate may be potential new targets for the prevention and treatment of liver fibrosis.