Merck
  • Leptin increases sympathetic nerve activity via induction of its own receptor in the paraventricular nucleus.
Leptin increases sympathetic nerve activity via induction of its own receptor in the paraventricular nucleus.
eLife (2020-06-17)
Zhigang Shi, Nicole E Pelletier, Jennifer Wong, Baoxin Li, Andrei D Sdrulla, Christopher J Madden, Daniel L Marks, Virginia L Brooks
ABSTRACT
Whether leptin acts in the paraventricular nucleus (PVN) to increase sympathetic nerve activity (SNA) is unclear, since PVN leptin receptors (LepR) are sparse. We show in rats that PVN leptin slowly increases SNA to muscle and brown adipose tissue, because it induces the expression of its own receptor and synergizes with local glutamatergic neurons. PVN LepR are not expressed in astroglia and rarely in microglia; instead, glutamatergic neurons express LepR, some of which project to a key presympathetic hub, the rostral ventrolateral medulla (RVLM). In PVN slices from mice expressing GCaMP6, leptin excites glutamatergic neurons. LepR are expressed mainly in thyrotropin-releasing hormone (TRH) neurons, some of which project to the RVLM. Injections of TRH into the RVLM and dorsomedial hypothalamus increase SNA, highlighting these nuclei as likely targets. We suggest that this neuropathway becomes important in obesity, in which elevated leptin maintains the hypothalamic pituitary thyroid axis, despite leptin resistance.
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Thyrotropin releasing hormone, ≥98% (HPLC), powder
Sigma-Aldrich
Goat serum