Merck
  • Lysine to arginine mutagenesis of chlorotoxin enhances its cellular uptake.
Lysine to arginine mutagenesis of chlorotoxin enhances its cellular uptake.
Biopolymers (2017-05-02)
Paola G Ojeda, Sónia Troeira Henriques, Yijun Pan, Joseph A Nicolazzo, David J Craik, Conan K Wang
ABSTRACT
Chlorotoxin (CTX), a disulfide-rich peptide from the scorpion Leiurus quinquestriatus, has several promising biopharmaceutical properties, including preferential affinity for certain cancer cells, high serum stability, and cell penetration. These properties underpin its potential for use as a drug design scaffold, especially for the treatment of cancer; indeed, several analogs of CTX have reached clinical trials. Here, we focus on its ability to internalize into cells-a trait associated with a privileged subclass of peptides called cell-penetrating peptides-and whether it can be improved through conservative substitutions. Mutants of CTX were made using solid-phase peptide synthesis and internalization into human cervical carcinoma (HeLa) cells was monitored by fluorescence and confocal microscopy. CTX_M1 (ie, [K15R/K23R]CTX) and CTX_M2 (ie, [K15R/K23R/Y29W]CTX) mutants showed at least a twofold improvement in uptake compared to CTX. We further showed that these mutants internalize into HeLa cells largely via an energy-dependent mechanism. Importantly, the mutants have high stability, remaining intact in serum for over 24 h; thus, retaining the characteristic stability of their parent peptide. Overall, we have shown that simple conservative substitutions can enhance the cellular uptake of CTX, suggesting that such type of mutations might be useful for improving uptake of other peptide toxins.
Sigma-Aldrich
Chlorotoxin from Leiurus quinquestriatus (north Africa), recombinant, expressed in E. coli, ≥98% (HPLC), lyophilized powder