Merck
  • Design and synthesis of 7-alkoxy-4-heteroarylamino-3-quinolinecarbonitriles as dual inhibitors of c-Src kinase and nitric oxide synthase.
Design and synthesis of 7-alkoxy-4-heteroarylamino-3-quinolinecarbonitriles as dual inhibitors of c-Src kinase and nitric oxide synthase.
Bioorganic & medicinal chemistry (2008-05-20)
Xin Cao, Qi-Dong You, Zhi-Yu Li, Qing-Long Guo, Jing Shang, Ming Yan, Ji-Wang Chern, Men-Ling Chen
ABSTRACT
Because both c-Src and iNOS are key regulatory enzymes in tumorigenesis, a new series of 4-heteroarylamino-3-quinolinecarbonitriles as potent dual inhibitors of both enzymes were designed, prepared, and evaluated for blocking multiple signaling pathways in cancer therapy. All compounds were evaluated by two related enzyme inhibition assays and an anti-proliferation assay in vitro. The results showed that most compounds could inhibit both enzymes, and several of them showed potent inhibition activity against different cancer cell lines. The best compound 20 (CPU-Y020) showed the IC(50) values of 6.58 and 7.61microM toward colon cancer HT-29 and liver cancer HepG2 cell lines.
Sigma-Aldrich
2-Aminothiazole, 97%