Merck
  • Activation and enzymatic characterization of recombinant human kallikrein 8.
Activation and enzymatic characterization of recombinant human kallikrein 8.
Biological chemistry (2006-06-28)
Tadaaki Kishi, Sylvain M Cloutier, Christoph Kündig, David Deperthes, Eleftherios P Diamandis
ABSTRACT
Human kallikrein 8 (hK8), whose gene was originally cloned as the human ortholog of a mouse brain protease, is known to be associated with diseases such as ovarian cancer and Alzheimer's disease. Recombinant human pro-kallikrein 8 was activated with lysyl endopeptidase-conjugated beads. Amino-terminal sequencing of the activated enzyme demonstrated the cleavage of a 9-aa propeptide from the pro-enzyme. The substrate specificity of activated hK8 was characterized using synthetic fluorescent substrates. hK8 showed trypsin-like specificity, as predicted from sequence analysis and enzymatic characterization of the mouse ortholog. All synthetic substrates tested containing either arginine or lysine at P1 position were cleaved by hK8. The highest kcat/Km value of 20x10(3)M-1 s-1 was observed with Boc-Val-Pro-Arg-7-amido-4-methylcoumarin. The activity of hK8 was inhibited by antipain, chymostatin, and leupeptin. The concentration for 50% inhibition by the best inhibitor, antipain, was 0.46 microM. The effect of different metal ions on the enzyme activity was analyzed. Whereas Na+ had no effect on hK8 activity, Ni2+ and Zn2+ decreased the activity and Ca2+, Mg2+, and K+ had a stimulatory effect. Ca2+ was the best activator, with an optimal concentration of approximately 10 microM.
Sigma-Aldrich
Boc-Val-Pro-Arg-7-amido-4-methylcoumarin hydrochloride
Pricing and availability is not currently available.