- Sterilization of MacConkey agar and CLED medium by gamma-radiation.
Sterilization of MacConkey agar and CLED medium by gamma-radiation.
The International journal of applied radiation and isotopes (1983-10-01)
B Bogokowsky, E Eisenberg, G Altmann
PMID6358048
ABSTRACT
MacConkey agar and Cystine-Lactose-Electrolyte-Deficient (CLED) agar, media widely used in the bacteriological laboratory and recommended for the detection of urinary tract infections, were sterilized by gamma-radiation at a dose of 1.5 Mrad. Both were modified and adapted to radiation sterilization by adding sodium thioglycollate as a radioprotectant, and by increasing their indicator content. The media performed well when tested with different Enterobacteria and other micro-organisms. Growth and change of indicator reaction were equal in irradiated and autoclaved culture media. Culture media were also evaluated after storage for one month at room temperature and at 4 degrees C and compared well with freshly autoclaved media.