Merck
  • Oncogenic EP300 can be targeted with inhibitors of aldo-keto reductases.
Oncogenic EP300 can be targeted with inhibitors of aldo-keto reductases.
Biochemical pharmacology (2019-03-14)
Zimam Mahmud, Muhammad Asaduzzaman, Uttom Kumar, Nahal Masrour, Roman Jugov, R Charles Coombes, Sami Shousha, Yunhui Hu, Eric W-F Lam, Ernesto Yagüe
ABSTRACT
E-cadherin transcriptional activator EP300 is down-regulated in metaplastic breast carcinoma, a rare form of triple negative and E-cadherin-negative aggressive breast cancer with a poor clinical outcome. In order to shed light on the regulation of E-cadherin by EP300 in breast cancer we analyzed by immunohistochemistry 41 cases of invasive breast cancer with both E-cadherinhigh and E-cadherinlow expression levels, together with 20 non-malignant breast tissues. EP300 and E-cadherin showed a positive correlation in both non-malignant and cancer cases and both markers together were better predictors of lymph node metastasis than E-cadherin alone. These data support a metastasis suppressor role for EP300 in breast cancer. However, some reports suggest an oncogenic role for EP300. We generated a breast cancer cell model to study E-cadherin-independent effects of EP300 by over-expression of EP300 in HS578T cells which have E-cadherin promoter hypermethylated. In this cell system, EP300 led to up-regulation of mesenchymal (vimentin, Snail, Slug, Zeb1) and stemness (ALDH+ and CD44high/CD24low) markers, increases in migration, invasion, anchorage-independent growth and drug resistance. Genome-wide expression profiling identified aldo-keto reductases AKR1C1-3 as effectors of stemness and drug resistance, since their pharmacological inhibition with flufenamic acid restored both doxorubicin and paclitaxel sensitivity and diminished mammosphere formation. Thus, in cells with a permissive E-cadherin promoter, EP300 acts as a tumour/metastasis supressor by up-regulating E-cadherin expression, maintenance of the epithelial phenotype and avoidance of an epithelial-to-mesenchymal transition. In cells in which the E-cadherin promoter is hypermethylated, EP300 functions as an oncogene via up-regulation of aldo-keto reductases. This offers the rationale of using current aldo-keto reductase inhibitors in breast cancer treatment.
Sigma-Aldrich
Anti-EP300 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution