Merck
  • Endophilin A2 deficiency protects rodents from autoimmune arthritis by modulating T cell activation.
Endophilin A2 deficiency protects rodents from autoimmune arthritis by modulating T cell activation.
Nature communications (2021-01-29)
Ulrika Norin, Carola Rintisch, Liesu Meng, Florian Forster, Diana Ekman, Jonatan Tuncel, Katrin Klocke, Johan Bäcklund, Min Yang, Michael Y Bonner, Gonzalo Fernandez Lahore, Jaime James, Klementy Shchetynsky, Maria Bergquist, Inger Gjertsson, Norbert Hubner, Liselotte Bäckdahl, Rikard Holmdahl
ABSTRACT
The introduction of the CTLA-4 recombinant fusion protein has demonstrated therapeutic effects by selectively modulating T-cell activation in rheumatoid arthritis. Here we show, using a forward genetic approach, that a mutation in the SH3gl1 gene encoding the endocytic protein Endophilin A2 is associated with the development of arthritis in rodents. Defective expression of SH3gl1 affects T cell effector functions and alters the activation threshold of autoreactive T cells, thereby leading to complete protection from chronic autoimmune inflammatory disease in both mice and rats. We further show that SH3GL1 regulates human T cell signaling and T cell receptor internalization, and its expression is upregulated in rheumatoid arthritis patients. Collectively our data identify SH3GL1 as a key regulator of T cell activation, and as a potential target for treatment of autoimmune diseases.
Sigma-Aldrich
Anti-SH3GL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution