Merck
  • Synthesis and screening of mono- and di-aryl technetium and rhenium metallocarboranes. A new class of probes for the estrogen receptor.
Synthesis and screening of mono- and di-aryl technetium and rhenium metallocarboranes. A new class of probes for the estrogen receptor.
Journal of medicinal chemistry (2008-04-17)
Patrick W Causey, Travis R Besanger, John F Valliant
ABSTRACT
A series of mono and diaryl rhenium(I)-carborane derivatives were prepared using microwave heating and screened for their affinity for two isoforms of the estrogen receptor (ER). The rhenacarborane derivative [(RR'C 2B9H9)Re(CO)3](-) (R = p-PhOH, R' = H), which was generated by taking advantage of a recently discovered cage isomerization process, and the neutral nitrosated analogue [(RR'C2B9H9)Re(CO)2(NO)] (R = p-PhOH, R' = H) showed the highest affinities of the compounds screened. As a result, the (99m)Tc analogue of one of the leads was produced in high yield (84%) and specific activity in a manner that is suitable for routine production in support of future preclinical and molecular imaging studies.
Product Description
Supelco
Pentachlorophenol solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
o-Cresol, ≥99%
Sigma-Aldrich
Pentachlorophenol, 97%
Sigma-Aldrich
4-Phenylphenol, 97%
Sigma-Aldrich
p-Cresol, ≥99%, FG
Sigma-Aldrich
4-Phenylphenol, purified by sublimation, 99%
Sigma-Aldrich
4-Chlorophenol, ≥99%
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Supelco
o-Cresol, analytical standard
Supelco
p-Cresol, analytical standard
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
o-Cresol, ReagentPlus®, ≥99%
Sigma-Aldrich
p-Cresol, 99%
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Diphenyl ether, ReagentPlus®, 99%
Sigma-Aldrich
Diphenyl ether, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Diphenyl ether, ≥99%, FG
Sigma-Aldrich
2,4,6-Trichlorophenol, 98%
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology