Merck
  • Comprehensive Comparison of Two Color Varieties of Perillae Folium Using Rapid Resolution Liquid Chromatography Coupled with Quadruple-Time-of-Flight Mass Spectrometry (RRLC-Q/TOF-MS)-Based Metabolic Profile and in Vivo/in Vitro Anti-Oxidative Activity.
Comprehensive Comparison of Two Color Varieties of Perillae Folium Using Rapid Resolution Liquid Chromatography Coupled with Quadruple-Time-of-Flight Mass Spectrometry (RRLC-Q/TOF-MS)-Based Metabolic Profile and in Vivo/in Vitro Anti-Oxidative Activity.
Journal of agricultural and food chemistry (2020-11-26)
Yun-Feng Zheng, Dan-Yang Li, Jie Sun, Jian-Ming Cheng, Chuan Chai, Li Zhang, Guo-Ping Peng
ABSTRACT
Perillae Folium (PF), which is extensively used as a dietary vegetable and medicinal herb, contains two varietal forms corresponding to purple perilla leaf (Perilla frutescens var. crispa) and green perilla leaf (Perilla frutescens var. frutescens). However, the components and efficacy of different PF varieties remain underexplored so far. In the present work, a nontargeted rapid resolution liquid chromatography coupled with quadruple-time-of-flight mass spectrometry (RRLC-Q/TOF-MS)-based metabolomics approach was developed to investigate the difference in the chemical compositions between green PF and purple PF. A total of 71 compounds were identified or tentatively identified, among which 7 phenolic acids, 10 flavonoids, and 9 anthocyanins were characterized as differential metabolites. In addition, heatmap visualization and ultraperformance liquid chromatography coupled with triple-quadrupole tandem mass spectrometry (UPLC-TQ-MS/MS)-based quantitative analysis revealed that flavonoids and anthocyanins especially had higher contents in purple PF. Furthermore, the anti-oxidative activities of two varietal PFs were evaluated in vivo zebrafish and in vitro human umbilical vein endothelial cells (HUVECs). The results showed that the purple PF had more pronounced anti-oxidative activities than did the green PF, which may be due to the presence of anthocyanins and a higher concentration of flavonoids in its phytochemical profile. The outcome of the present study is expected to provide useful insight on the comprehensive utilization of a PF resource.
Sigma-Aldrich
Acridine Orange hydrochloride solution, ≥95.0% (HPLC), 10 mg/mL in H2O
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile