Merck
  • Synthesis and anti-inflammatory/antioxidant activities of some new ring substituted 3-phenyl-1-(1,4-di-N-oxide quinoxalin-2-yl)-2-propen-1-one derivatives and of their 4,5-dihydro-(1H)-pyrazole analogues.
Synthesis and anti-inflammatory/antioxidant activities of some new ring substituted 3-phenyl-1-(1,4-di-N-oxide quinoxalin-2-yl)-2-propen-1-one derivatives and of their 4,5-dihydro-(1H)-pyrazole analogues.
Bioorganic & medicinal chemistry letters (2007-10-19)
Asunción Burguete, Eleni Pontiki, Dimitra Hadjipavlou-Litina, Raquel Villar, Esther Vicente, Beatriz Solano, Saioa Ancizu, Silvia Pérez-Silanes, Ignacio Aldana, Antonio Monge
ABSTRACT
We report the synthesis, anti-inflammatory and antioxidant activities of novel ring substituted 3-phenyl-1-(1,4-di-N-oxide quinoxalin-2-yl)-2-propen-1-one derivatives and of their 4,5-dihydro-(1H)-pyrazole analogues. The tested compounds inhibit the carrageenin-induced rat paw edema (4.5-56.1%) and present important scavenging activities. Compound 2a is the most potent (56.1%) in the in vivo experiment and exhibits promising in vitro inhibition of soybean lipoxygenase (IC(50)<1microM).
Sigma-Aldrich
Nordihydroguaiaretic acid, ≥97.0% (HPLC)
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Supelco
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)