Merck
  • Carbonic anhydrase inhibitors: the beta-carbonic anhydrase from Helicobacter pylori is a new target for sulfonamide and sulfamate inhibitors.
Carbonic anhydrase inhibitors: the beta-carbonic anhydrase from Helicobacter pylori is a new target for sulfonamide and sulfamate inhibitors.
Bioorganic & medicinal chemistry letters (2007-05-08)
Isao Nishimori, Tomoko Minakuchi, Takuhiro Kohsaki, Saburo Onishi, Hiroaki Takeuchi, Daniela Vullo, Andrea Scozzafava, Claudiu T Supuran
ABSTRACT
DNA clones for the beta-class carbonic anhydrase (CA, EC 4.2.1.1) of Helicobactor pylori (hpbetaCA) were obtained. A recombinant hpbetaCA protein lacking the N-terminal 15-amino acid residues was produced and purified, representing a catalytically efficient CA. hpbetaCA was strongly inhibited (K(I)s in the range of 24-45 nM) by many sulfonamides/sulfamates, among which acetazolamide, ethoxzolamide, topiramate, and sulpiride, all clinically used drugs. The dual inhibition of alpha- and/or beta-class CAs of H. pylori might represent a useful alternative for the management of gastritis/gastric ulcers, as well as gastric cancer. This is also the first study showing that a bacterial beta-CA can be a drug target.
Sigma-Aldrich
Sulfanilamide, ≥98%
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Sigma-Aldrich
4-Sulfamoylbenzoic acid, 97%
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Sigma-Aldrich
(±)-Sulpiride
Sigma-Aldrich
p-Toluenesulfonamide, ReagentPlus®, ≥99%
Sigma-Aldrich
p-Toluenesulfonamide, reagent grade, 97%