Merck
  • TSPO ligand etifoxine attenuates LPS-induced cognitive dysfunction in mice.
TSPO ligand etifoxine attenuates LPS-induced cognitive dysfunction in mice.
Brain research bulletin (2020-10-20)
Hui Zhang, Li Ma, Wen-Zhi Guo, Lin-Bo Jiao, Hong-Yu Zhao, Ya-Qun Ma, Xue-Mei Hao
ABSTRACT
The translocator protein (TSPO), once known as peripheral-type benzodiazepine receptor, was reported to be related with several physiological functions. Etifoxine is a clinically available anxiolytic drug, and has recently shown neuroprotective effects as a TSPO ligand. The aim of the present study was to investigate the influence of etifoxine on LPS-induced neuroinflammation and cognitive dysfunction. C57/BL6 male mice were injected with etifoxine (50 mg/kg, i.p.) three days before lipopolysaccharide (LPS, 500 μg/kg, i.p.) administration. Etifoxine pretreatment alleviated hippocampal inflammation, increased brain levels of progesterone, allopregnanolone and attenuated cognitive dysfunction in LPS-injected mice. While LPS increased expression of caspase-3 and decreased p-Akt/Akt, etifoxine returned caspase-3 and p-Akt/Akt to control levels. Finasteride, a 5α-reductase inhibitor that blocked allopregnanolone production, partially reversed the effects of etifoxine. We concluded that etifoxine exerted neuroprotective effects in LPS-induced neuroinflammation and the neuroprotection may be related with increase of neurosteroids synthesis and decrease of apoptosis.
Sigma-Aldrich
Etifoxine hydrochloride, ≥98% (HPLC)