Merck
  • Novel 3-O-glycosyl-3-demethylthiocolchicines as ligands for glycine and gamma-aminobutyric acid receptors.
Novel 3-O-glycosyl-3-demethylthiocolchicines as ligands for glycine and gamma-aminobutyric acid receptors.
Journal of medicinal chemistry (2007-04-07)
Maria Luisa Gelmi, Gabriele Fontana, Donato Pocar, Guido Pontremoli, Sara Pellegrino, Ezio Bombardelli, Antonella Riva, Walter Balduini, Silvia Carloni, Mauro Cimino
ABSTRACT
New 3-O-glycosyl-3-demethylthiocolchicines containing natural and unnatural sugar moieties were prepared and tested on gamma-aminobutyric acid (GABA) and strychnine-sensitive glycine receptors present in rat brain and spinal cord. Two different synthetic approaches were used with the readily available 3-O-demethylthiocolchicine (1b) and thiocolchicoside (2a). Glycosyl compounds 2a-g were obtained from 1b and 1-fluorosugars 4. 6'-Heterosubstituted glycosyl compounds 6-12 and the 6'-desoxy derivative 2h were prepared from 2a.
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
SAFC
Glycine
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine 1 M solution