Merck
  • The RNA helicase DDX5 supports mitochondrial function in small cell lung cancer.
The RNA helicase DDX5 supports mitochondrial function in small cell lung cancer.
The Journal of biological chemistry (2020-05-08)
Zheng Xing, Matthew P Russon, Sagar M Utturkar, Elizabeth J Tran, Zheng Xing, Matthew P Russon, Sagar M Utturkar, Elizabeth J Tran
ABSTRACT
DEAD-box helicase 5 (DDX5) is a founding member of the DEAD-box RNA helicase family, a group of enzymes that regulate ribonucleoprotein formation and function in every aspect of RNA metabolism, ranging from synthesis to decay. Our laboratory previously found that DDX5 is involved in energy homeostasis, a process that is altered in many cancers. Small cell lung cancer (SCLC) is an understudied cancer type for which effective treatments are currently unavailable. Using an array of methods, including short hairpin RNA-mediated gene silencing, RNA and ChIP sequencing analyses, and metabolite profiling, we show here that DDX5 is overexpressed in SCLC cell lines and that its down-regulation results in various metabolic and cellular alterations. Depletion of DDX5 resulted in reduced growth and mitochondrial dysfunction in the chemoresistant SCLC cell line H69AR. The latter was evidenced by down-regulation of genes involved in oxidative phosphorylation and by impaired oxygen consumption. Interestingly, DDX5 depletion specifically reduced intracellular succinate, a TCA cycle intermediate that serves as a direct electron donor to mitochondrial complex II. We propose that the oncogenic role of DDX5, at least in part, manifests as up-regulation of respiration supporting the energy demands of cancer cells.
Sigma-Aldrich
Anti-p68 Antibody, clone204, clone 204, Upstate®, from mouse
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid