Merck
  • Synthesis and pharmacological evaluation of a selected library of new potential anti-inflammatory agents bearing the gamma-hydroxybutenolide scaffold: a new class of inhibitors of prostanoid production through the selective modulation of microsomal prostaglandin E synthase-1 expression.
Synthesis and pharmacological evaluation of a selected library of new potential anti-inflammatory agents bearing the gamma-hydroxybutenolide scaffold: a new class of inhibitors of prostanoid production through the selective modulation of microsomal prostaglandin E synthase-1 expression.
Journal of medicinal chemistry (2007-04-05)
Maria D Guerrero, Maurizio Aquino, Ines Bruno, María C Terencio, Miguel Paya, Raffaele Riccio, Luigi Gomez-Paloma
ABSTRACT
As a part of our drug discovery effort, recently we clarified the molecular basis of phospholipase A2 (PLA2) inactivation by petrosaspongiolide M (PM), an interesting metabolite belonging to a marine sesterterpene family, containing in its structural architecture a gamma-hydroxybutenolide moiety and showing potent anti-inflammatory activity. In the attempt to expand structural diversity as well as to simplify crucial synthetic features of the parent compound, we decided to develop a selected library based on the densely functionalized gamma-hydroxybutenolide scaffold. The synthesized products were tested for their ability to inhibit PLA2 enzymes as well as to modulate the expression of inducible cyclooxygenase 2 (COX-2) and microsomal prostaglandin E synthase 1 (mPGES-1), two key enzymes highly involved in the inflammatory event, in order to discover new promising anti-inflammatory agents with better pharmacological profiles. This led us to the discovery of a promising inhibitor (4e) of prostanoid production acting by in vitro and in vivo selective modulation of microsomal prostaglandin E synthase 1 expression.
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
CAY10526, ≥98% (HPLC)