Merck
  • Actin microfilament aggregation induced by withaferin A is mediated by annexin II.
Actin microfilament aggregation induced by withaferin A is mediated by annexin II.
Nature chemical biology (2006-01-13)
Ryan R Falsey, Marilyn T Marron, G M Kamal B Gunaherath, Nikhil Shirahatti, Daruka Mahadevan, A A Leslie Gunatilaka, Luke Whitesell
ABSTRACT
The actin cytoskeleton supports diverse cellular processes such as endocytosis, oriented growth, adhesion and migration. The dynamic nature of the cytoskeleton, however, has made it difficult to define the roles of the many accessory molecules that modulate actin organization, especially the multifunctional adapter protein annexin II. We now report that the compound withaferin A (1) can alter cytoskeletal architecture in a previously unknown manner by covalently binding annexin II and stimulating its basal F-actin cross-linking activity. Drug-mediated disruption of F-actin organization is dependent on annexin II expression by cells and markedly limits their migratory and invasive capabilities at subcytotoxic concentrations. Given the extensive ethnobotanical history of withaferin-containing plant preparations in the treatment of cancer and inflammatory and neurological disorders, we suggest that annexin II represents a feasible, previously unexploited target for therapeutic intervention by small-molecule drugs.
Sigma-Aldrich
N-Ethylmaleimide, BioXtra, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
N-Ethylmaleimide, purum p.a., ≥99.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
N-Ethylmaleimide, crystalline, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Withaferin A, ≥95% (HPLC)
Pricing and availability is not currently available.