Merck
  • Patients with Essential Thrombocythemia may be Poor Responders to Enteric-Coated Aspirin, but not to Plain Aspirin.
Patients with Essential Thrombocythemia may be Poor Responders to Enteric-Coated Aspirin, but not to Plain Aspirin.
Thrombosis and haemostasis (2020-07-28)
Mariangela Scavone, Jessica Rizzo, Eti A Femia, Gian Marco Podda, Elena Bossi, Sabrina Caberlon, Rita Paroni, Marco Cattaneo
ABSTRACT
Essential thrombocythemia (ET) patients are treated with aspirin (acetylsalicylic acid [ASA]) to prevent thrombosis. Previous studies showed that serum thromboxane (Tx) B2 was high 24 hours after enteric-coated (EC)-ASA in ET patients, due to increased number of noninhibited reticulated platelets (RPs), consequent to high platelet turnover, and that ASA should be given twice a day to ET patients. We studied ET patients (n = 17) and healthy subjects (n = 10) on 100 mg EC-ASA once daily; experiments were repeated after 14-day treatment with 100 mg plain-ASA once daily. Serum TxB2, plasma ASA, and salicylic acid (SA) were measured before the morning dose and up to 8 hours thereafter. Blood activity of ASA-deacethylating esterases, in vitro inhibition of collagen-induced TxB2 production by ASA (10-1,000 µM), and number of RP were measured. TxB2 inhibition by ASA in vitro and esterases activities were normal in all subjects. EC-ASA elicited highly variable responses; 6 ET patients were poor responders, as their serum TxB2 was high after EC-ASA; their plasma levels of ASA and SA were low/undetectable. In contrast to EC-ASA, plain ASA decreased serum TxB2 and increased plasma ASA and SA in all subjects. Serum TxB2 was high in ET patients at 24 hours and significantly correlated with RP count (but not RP percentage) and platelet count. Plain ASA should be used in ET patients to inhibit platelets efficiently. The identification of ET patients who might benefit from twice a day ASA could simply be based on their platelet count: since their platelet turnover is not increased, ET patients with normalized platelet count should not need twice a day ASA treatment.
Sigma-Aldrich
lysine acetylsalicylic acid, AldrichCPR
Sigma-Aldrich
Thiazole Orange, Dye content ~90 %