Merck
  • Comprehensive Multi-omics Analysis Reveals Mitochondrial Stress as a Central Biological Hub for Spaceflight Impact.
Comprehensive Multi-omics Analysis Reveals Mitochondrial Stress as a Central Biological Hub for Spaceflight Impact.
Cell (2020-11-27)
Willian A da Silveira, Hossein Fazelinia, Sara Brin Rosenthal, Evagelia C Laiakis, Man S Kim, Cem Meydan, Yared Kidane, Komal S Rathi, Scott M Smith, Benjamin Stear, Yue Ying, Yuanchao Zhang, Jonathan Foox, Susana Zanello, Brian Crucian, Dong Wang, Adrienne Nugent, Helio A Costa, Sara R Zwart, Sonja Schrepfer, R A Leo Elworth, Nicolae Sapoval, Todd Treangen, Matthew MacKay, Nandan S Gokhale, Stacy M Horner, Larry N Singh, Douglas C Wallace, Jeffrey S Willey, Jonathan C Schisler, Robert Meller, J Tyson McDonald, Kathleen M Fisch, Gary Hardiman, Deanne Taylor, Christopher E Mason, Sylvain V Costes, Afshin Beheshti, Willian A da Silveira, Hossein Fazelinia, Sara Brin Rosenthal, Evagelia C Laiakis, Man S Kim, Cem Meydan, Yared Kidane, Komal S Rathi, Scott M Smith, Benjamin Stear, Yue Ying, Yuanchao Zhang, Jonathan Foox, Susana Zanello, Brian Crucian, Dong Wang, Adrienne Nugent, Helio A Costa, Sara R Zwart, Sonja Schrepfer, R A Leo Elworth, Nicolae Sapoval, Todd Treangen, Matthew MacKay, Nandan S Gokhale, Stacy M Horner, Larry N Singh, Douglas C Wallace, Jeffrey S Willey, Jonathan C Schisler, Robert Meller, J Tyson McDonald, Kathleen M Fisch, Gary Hardiman, Deanne Taylor, Christopher E Mason, Sylvain V Costes, Afshin Beheshti
ABSTRACT
Spaceflight is known to impose changes on human physiology with unknown molecular etiologies. To reveal these causes, we used a multi-omics, systems biology analytical approach using biomedical profiles from fifty-nine astronauts and data from NASA's GeneLab derived from hundreds of samples flown in space to determine transcriptomic, proteomic, metabolomic, and epigenetic responses to spaceflight. Overall pathway analyses on the multi-omics datasets showed significant enrichment for mitochondrial processes, as well as innate immunity, chronic inflammation, cell cycle, circadian rhythm, and olfactory functions. Importantly, NASA's Twin Study provided a platform to confirm several of our principal findings. Evidence of altered mitochondrial function and DNA damage was also found in the urine and blood metabolic data compiled from the astronaut cohort and NASA Twin Study data, indicating mitochondrial stress as a consistent phenotype of spaceflight.
Sigma-Aldrich
4-Nitrobenzoic acid, purum, ≥98.0% (HPLC)
Sigma-Aldrich
8-Cyclopentyl-1,3-dimethylxanthine, ≥98% (HPLC), powder
Supelco
Myristoyl-L-carnitine, analytical standard
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
Val-Tyr-Val, crystalline
Supelco
Chlorpropamide, analytical standard, ≥97%
Sigma-Aldrich
L-Carnitine hydrochloride, synthetic, ≥98%
Sigma-Aldrich
Debrisoquine sulfate
Sigma-Aldrich
Benzyldimethyldodecylammonium chloride, ≥99.0% (AT)
Supelco
D-Malic acid, analytical standard
Debrisoquine sulfate, British Pharmacopoeia (BP) Reference Standard