Merck
  • Modeling K(m) values using electrotopological state: substrates for cytochrome P450 3A4-mediated metabolism.
Modeling K(m) values using electrotopological state: substrates for cytochrome P450 3A4-mediated metabolism.
Bioorganic & medicinal chemistry letters (2005-07-02)
Yong-Hua Wang, Yan Li, Yan-Hong Li, Sheng-Li Yang, Ling Yang
ABSTRACT
In order to determine K(m) values of substrates for CYP3A4-mediated metabolism, an in silico model has been developed in the present work. Using electrotopological state (E-state) indices, together with Bayesian-regularized neural network (BRNN), we have described an in silico method to model log(1/K(m)) values of various substrates. The relative importance of the E-state indices is analyzed by principal component analysis. By using an additional external test set, which is independent of the training set, the robustness and predictivity of the model are also validated.
Product Description
Supelco
Dexamethasone, VETRANAL®, analytical standard
Supelco
Dapsone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Quinine, suitable for fluorescence, anhydrous, ≥98.0% (dried material, NT)
Sigma-Aldrich
Quinine, 90%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sigma-Aldrich
Terfenadine
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Colchicine, BioReagent, suitable for plant cell culture, ≥95% (HPLC)
Sigma-Aldrich
Bromodichloromethane, ≥97%
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, for molecular biology, ≥95%
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sigma-Aldrich
7-Ethoxycoumarin
Sigma-Aldrich
Hydrocortisone-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Carbamazepine, powder