Merck
  • Eomes-Dependent Loss of the Co-activating Receptor CD226 Restrains CD8+ T Cell Anti-tumor Functions and Limits the Efficacy of Cancer Immunotherapy.
Eomes-Dependent Loss of the Co-activating Receptor CD226 Restrains CD8+ T Cell Anti-tumor Functions and Limits the Efficacy of Cancer Immunotherapy.
Immunity (2020-10-15)
Marianne Weulersse, Assia Asrir, Andrea C Pichler, Lea Lemaitre, Matthias Braun, Nadège Carrié, Marie-Véronique Joubert, Marie Le Moine, Laura Do Souto, Guillaume Gaud, Indrajit Das, Elisa Brauns, Clara M Scarlata, Elena Morandi, Ashmitha Sundarrajan, Marine Cuisinier, Laure Buisson, Sabrina Maheo, Sahar Kassem, Arantxa Agesta, Michaël Pérès, Els Verhoeyen, Alejandra Martinez, Julien Mazieres, Loïc Dupré, Thomas Gossye, Vera Pancaldi, Camille Guillerey, Maha Ayyoub, Anne S Dejean, Abdelhadi Saoudi, Stanislas Goriely, Hervé Avet-Loiseau, Tobias Bald, Mark J Smyth, Ludovic Martinet
ABSTRACT
CD8+ T cells within the tumor microenvironment (TME) are exposed to various signals that ultimately determine functional outcomes. Here, we examined the role of the co-activating receptor CD226 (DNAM-1) in CD8+ T cell function. The absence of CD226 expression identified a subset of dysfunctional CD8+ T cells present in peripheral blood of healthy individuals. These cells exhibited reduced LFA-1 activation, altered TCR signaling, and a distinct transcriptomic program upon stimulation. CD226neg CD8+ T cells accumulated in human and mouse tumors of diverse origin through an antigen-specific mechanism involving the transcriptional regulator Eomesodermin (Eomes). Despite similar expression of co-inhibitory receptors, CD8+ tumor-infiltrating lymphocyte failed to respond to anti-PD-1 in the absence of CD226. Immune checkpoint blockade efficacy was hampered in Cd226-/- mice. Anti-CD137 (4-1BB) agonists also stimulated Eomes-dependent CD226 loss that limited the anti-tumor efficacy of this treatment. Thus, CD226 loss restrains CD8+ T cell function and limits the efficacy of cancer immunotherapy.
Sigma-Aldrich
Sodium chloride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Trizma® hydrochloride solution, pH 7.5, BioPerformance Certified, 1 M, suitable for cell culture