Merck
An ErbB2 splice variant lacking exon 16 drives lung carcinoma.
Proceedings of the National Academy of Sciences of the United States of America (2020-07-31)
Harvey W Smith, Lei Yang, Chen Ling, Arlan Walsh, Victor D Martinez, Jonathan Boucher, Dongmei Zuo, Ethan S Sokol, Dean C Pavlick, Garrett M Frampton, Juliann Chmielecki, Laura M Jones, Philippe P Roux, William W Lockwood, William J Muller
ABSTRACT
Lung cancer causes more deaths annually than any other malignancy. A subset of non-small cell lung cancer (NSCLC) is driven by amplification and overexpression or activating mutation of the receptor tyrosine kinase (RTK) ERBB2 In some contexts, notably breast cancer, alternative splicing of ERBB2 causes skipping of exon 16, leading to the expression of an oncogenic ERBB2 isoform (ERBB2ΔEx16) that forms constitutively active homodimers. However, the broader implications of ERBB2 alternative splicing in human cancers have not been explored. Here, we have used genomic and transcriptomic analysis to identify elevated ERBB2ΔEx16 expression in a subset of NSCLC cases, as well as splicing site mutations facilitating exon 16 skipping and deletions of exon 16 in a subset of these lung tumors and in a number of other carcinomas. Supporting the potential of ERBB2ΔEx16 as a lung cancer driver, its expression transformed immortalized lung epithelial cells while a transgenic model featuring inducible ERBB2ΔEx16 specifically in the lung epithelium rapidly developed lung adenocarcinomas following transgene induction. Collectively, these observations indicate that ERBB2ΔEx16 is a lung cancer oncogene with potential clinical importance for a proportion of patients.
Sigma-Aldrich
Hexadimethrine bromide, ≥95%
Sigma-Aldrich
Anti-erbB-2/HER-2 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
β-Glycerophosphate disodium salt hydrate, ≤1.0 mol % L-α-isomer
Sigma-Aldrich
Anti-Clara Cell Secretory Protein Antibody, serum, Upstate®